missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-60951
This item is not returnable.
View return policy
Description
CACNA2D1 Polyclonal specifically detects CACNA2D1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CACNA2D1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1 | |
| Goat | |
| 125 kDa | |
| 0.1 mg | |
| Primary | |
| Human Predicted cross-reactivity based on sequence identity: Dog, Pig, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| P54289 | |
| CACNA2D1 | |
| Peptide with sequence C-QEIFNKYNKDKKVR corresponding to internal region according to NP_000713.2. | |
| Affinity Purified | |
| RUO | |
| 781 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction