missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 381.00 - € 529.00
Specifications
| Antigen | CACNA2D2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216234
|
Novus Biologicals
NBP2-57784 |
100 μL |
€ 529.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640679
|
Novus Biologicals
NBP2-57784-25ul |
25 μL |
€ 381.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNA2D2 Polyclonal specifically detects CACNA2D2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CACNA2D2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 9254 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TNTNLLFVVAEKPLCSQCEAGRLLQKETHSDGPEQCELVQRPRYRRGPHICFDYNATEDTSDCGRGASFPPSLGVLVSLQLL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| alpha 2 delta calcium channel subunit, CACNA2D, calcium channel, voltage-dependent, alpha 2/delta subunit 2, gene 26, KIAA0558Voltage-gated calcium channel subunit alpha-2/delta-2, LUAC11.1, voltage-dependent calcium channel subunit alpha-2/delta-2 | |
| CACNA2D2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title