Learn More
Invitrogen™ CACNA2D3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587802
Description
Immunogen sequence: LYDYQAMCRA NKESSDGAHG LLDPYNAFLS AVKWIMTELV LFLVEFNLCS WWHSDMTAKA QKLKQTLEPC DTEYPAFVSE RTIKETTGNI ACEDCSKSFV IQQIPSSNLF MVVVDSSCLC ESVAPITMAP IEIRYNESLK C; Positive Samples: U-87 MG, HeLa, Hep G2, Mouse brain, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.
CACNA2D3 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits.
Specifications
| CACNA2D3 | |
| Polyclonal | |
| Unconjugated | |
| CACNA2D3 | |
| alpha 2 delta-3; alpha2delta3; Cacna2d3; Cacnad3; calcium channel alpha2-delta3 subunit; calcium channel, voltage dependent, alpha2/delta subunit 3; calcium channel, voltage-dependent, alpha 2/delta 3 subunit; calcium channel, voltage-dependent, alpha 2/delta subunit 3; calcium channel, voltage-dependent, alpha2/delta subunit 3; calcium voltage-gated channel auxiliary subunit alpha2delta 3; CaV alpha2delta3; HSA272268; Voltage-dependent calcium channel subunit alpha-2/delta-3; Voltage-dependent calcium channel subunit alpha-2-3; Voltage-dependent calcium channel subunit delta-3; Voltage-gated calcium channel subunit alpha-2/delta-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12294, 306243, 55799 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CFG5, Q8IZS8, Q9Z1L5 | |
| CACNA2D3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1040 of human CACNA2D3 (NP_0608682). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.