missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNA2D4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588418
This item is not returnable.
View return policy
Description
Immunogen sequence: AKSVFHHSHK HKKQDPLQPC DTEYPVFVYQ PAIREANGIV ECGPCQKVFV VQQIPNSNLL LLVTDPTCDC SIFPPVLQEA TEVKYNASVK CDRMRSQKLR RRPDSCHAFH PEENAQDCGG ASDT; Positive Samples: HT-29, LO2; Cellular Location: Membrane, Single-pass type I membrane protein.
CACNA2D4 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
| CACNA2D4 | |
| Polyclonal | |
| Unconjugated | |
| Cacna2d4 | |
| 5730412N02Rik; BE686333; Cacna2d4; calcium channel, voltage-dependent, alpha 2/delta subunit 4; calcium voltage-gated channel auxiliary subunit alpha2delta 4; LOW QUALITY PROTEIN: voltage-dependent calcium channel subunit alpha-2/delta-4; RCD4; RGD1562038; Voltage-dependent calcium channel subunit alpha-2/delta-4; Voltage-dependent calcium channel subunit alpha-2-4; Voltage-dependent calcium channel subunit delta-4; voltage-gated calcium channel alpha(2)delta-4 subunit; voltage-gated calcium channel subunit alpha-2/delta-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 93589 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.02 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7Z3S7 | |
| Cacna2d4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 992-1115 of human CACNA2D4 (NP_7589524). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction