missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNB2 Antibody (N8b/1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47607H
This item is not returnable.
View return policy
Description
CACNB2 Monoclonal specifically detects CACNB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CACNB2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CAB2, CACNLB2, Calcium channel voltage-dependent subunit beta 2, calcium channel, voltage-dependent, beta 2 subunit, CAVB2, FLJ23743, Lambert-Eaton myasthenic syndrome antigen B, myasthenic (Lambert-Eaton) syndrome antigen B, MYSB, voltage-dependent L-type calcium channel subunit beta-2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 78 kDa. No cross reactivity against CavB1, CavB3, CavB4. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S8b-1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q8VGC3 | |
| CACNB2 | |
| Synthetic peptide amino acids 189-205 of rat CavBeta2 | |
| 0.1 mL | |
| 783 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction