missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CACNB2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11596052
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11596052 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11596052 Supplier Invitrogen™ Supplier No. PA112669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for this product is rat brain lysate.

The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Cacnb2
Gene Accession No. Q08289
Gene Alias AW060387; brain calcium channel beta-subunit CaB2c (splice variant of the gene CaB2); CAB1; CAB2; Cacnb2; CACNLB1; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; calcium voltage-gated channel auxiliary subunit beta 2; cardiac calcium channel beta subunit; cardiac calcium channel beta-subunit CaB2a (splice variant of the gene CaB2; cardiac calcium channel beta-subunit CaB2b (splice variant of the gene CaB2); CAVB2; Cavbeta2; Cchb2; CCHLB1; lambert-Eaton myasthenic syndrome antigen B; L-type calcium channel beta subunit; L-type voltage-dependent calcium channel beta-2 subunit; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; voltage-dependent calcium channel beta2A subunit; voltage-dependent calcium channel beta2B subunit; voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Gene Symbols Cacnb2
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the rat beta2 calcium channel.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.