missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539527
This item is not returnable.
View return policy
Description
Voltage-dependent calcium channels are essential for the release of neurotransmitters. L-type (long lasting current) voltage-dependent calcium channels are composed of four subunits: an alpha1 subunit, a beta subunit, a gamma subunit and an alpha2 delta subunit. The beta subunit is encoded by four genes, designated beta1-beta4, all of which contribute to the diversity of calcium currents and are involved in membrane trafficking of the alpha1 subunit. L-type Ca ++ CP gamma1, also known as CACNLG or CACNG1, is a 222 amino acid multi-pass membrane protein belonging to the PMP-22/EMP/MP20 family. Expressed in skeletal muscle, L-type Ca ++ CP gamma1 is a subunit of the dihydropyridine (DHP) sensitive calcium channel and may play a role in excitation-contraction coupling. L-type Ca++ CP gamma1 is considered a novel marker for maligt hyperthermia susceptibility (MHS), an autosomal domit disorder of skeletal muscle which manifests as a life-threatening hypermetabolic crisis triggered by commonly used inhalation anaesthetics and depolarizing muscle relaxants.
Specifications
| CACNG1 | |
| Polyclonal | |
| Unconjugated | |
| CACNG1 | |
| Voltage-dependent calcium channel gamma-1 subunit, Dihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, CACNLG, L-type Ca++ CP gamma1, L-type Ca++ CP gamma1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12299, 29658, 786 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q06432, O70578, P97707 | |
| CACNG1 | |
| A synthetic peptide derived from the internal region of human CACNG1 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction