missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566769
This item is not returnable.
View return policy
Description
Immunogen sequence: TLSREPLKVTT AASYSPDQEA SFLQVHDFFQ QDLKEGFHVS MLNRRTTPV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
The expression of multiple classes of voltage dependent calcium channels (VDCC's) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC's exist before birth which is consistent with a role in migration, most N-Type VDCC's subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications
| CACNG4 | |
| Polyclonal | |
| Unconjugated | |
| Cacng4 | |
| AI413107; AW491861; CACNG4; calcium channel, voltage-dependent, gamma subunit 4; calcium voltage-gated channel auxiliary subunit gamma 4; CaV gamma4; MGC11138; MGC24983; neuronal voltage-gated calcium channel gamma-4 subunit; TARP gamma-4; transmembrane AMPAR regulatory protein gamma-4; Voltage-dependent calcium channel gamma-4 subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBN1 | |
| Cacng4 | |
| Recombinant Human Calcium Channel gamma-4. Recombinant protein control fragment (Product #RP-107426). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction