missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNG5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114706
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CACNG5.
L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.
Specifications
| CACNG5 | |
| Polyclonal | |
| Unconjugated | |
| Cacng5 | |
| Cacng5; calcium channel, voltage-dependent, gamma subunit 5; calcium voltage-gated channel auxiliary subunit gamma 5; neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; transmembrane AMPAR regulatory protein gamma-5; voltage dependent calcium channel gamma 5 subunit; voltage-dependent calcium channel gamma-5 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140723, 140726, 27091 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8VHW4, Q8VHW8, Q9UF02 | |
| Cacng5 | |
| A synthesized peptide derived from human CACNG5(Accession Q9UF02), corresponding to amino acid residues E67-L117. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction