Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
Product Code. 17614825
Change view
Click to view available options
Diameter (English) Ball:
0.074 in.
0.078 in.
Gauge:
18G
20G
Length (English):
1.2 in.
1.5 in.
2 in.
3 in.
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Ball Gauge Length (English) Unit Size
17614825 0.078 in. 18G 3 in. Pack of 100
16448213 0.078 in. 18G 3 in. Pack of 1000
15786118 0.074 in. 20G 1.2 in. Pack of 100
15282035 0.078 in. 18G 2 in. Pack of 100
15282065 0.078 in. 18G 1.5 in. Pack of 100
13244779 0.074 in. 20G 1.5 in. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17614825 Supplier Cadence Science™ Supplier No. 9924
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For critical applications requiring sterile, flexible, single-use feeding needles
  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box
Specifications
Product Type Animal Feeding Needle (Flexible)
Material PTFE
Disposable Single-use
For Use With (Application) For critical applications requiring sterile, flexible, single use feeding needles
Gauge 18G
Sterility Sterile
Length (English) 3 in.
Length (Metric) 0.75 cm
Diameter (English) Ball 0.078 in.
Diameter (Metric) Ball 0.02 cm