missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-12 Antibody (343621), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cadherin-12 Monoclonal antibody specifically detects Cadherin-12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
Specifications
| Antigen | Cadherin-12 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | 343621 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, FLJ34857, N-cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 |
| Host Species | Rat |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12, Met1-Pro609, Accession # P55289 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?