missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Antibody (OTI3H6), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70385PEATT594
This item is not returnable.
View return policy
Description
Cadherin-13 Monoclonal specifically detects Cadherin-13 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Cadherin-13 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CDH13 | |
| Full length recombinant protein of human Cadherin-13 (NP_001248) produced in HEK293T cells. | |
| 0.1 mL | |
| 1012.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI3H6 | |
| Flow Cytometry | |
| cadherin 13, H-cadherin (heart), cadherin-13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, Truncated cadherin, truncated-cadherin | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction