missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Antibody (03), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 335.00
Specifications
| Antigen | Cadherin-16 |
|---|---|
| Clone | 03 |
| Applications | ELISA, Sandwich ELISA Capture |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645828
|
Novus Biologicals
NBP3-06524-100ul |
100 μg |
€ 335.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616038
|
Novus Biologicals
NBP3-06524-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Sandwich ELISA CaptureSpecifications
| Cadherin-16 | |
| ELISA, Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| Human | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-16 (Accession#: NP_004053.1; Met1-Ala786). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 03 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 1014 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title