missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Antibody (CDH16/7028R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14214PE
This item is not returnable.
View return policy
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cadherin-16 | |
| Monoclonal | |
| PE | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| Recombinant human full-length Cadherin-16 protein (Uniprot: O75309 ) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CDH16/7028R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1014 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction