missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-59248
This item is not returnable.
View return policy
Description
Cadherin-16 Polyclonal specifically detects Cadherin-16 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cadherin-16 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| Rabbit | |
| 88 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| O75309 | |
| CDH16 | |
| Synthetic peptides corresponding to CDH16(cadherin 16, KSP-cadherin) The peptide sequence was selected from the N terminal of CDH16. Peptide sequence SDRDEPGTANSDLRFHILSQAPAQPSPDMFQLEPRLGALALSPKGSTSLD. | |
| Affinity purified | |
| RUO | |
| 1014 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion