missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Antibody (rCDH16/1071), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08881C
This item is not returnable.
View return policy
Description
Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| Cadherin-16 | |
| Monoclonal | |
| DyLight 650 | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| Recombinant full-length human Cadherin-16 protein (Uniprot: O75309) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| rCDH16/1071 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 1014 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction