missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-17 Antibody (rCDH17/8514), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20889AF488
This item is not returnable.
View return policy
Description
Cadherin-17 Monoclonal antibody specifically detects Cadherin-17 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cadherin-17 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
| Recombinant fragment (around aa 242-418) of human Cadherin-17 protein (CDH17) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 1015 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCDH17/8514 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction