missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-78811
This item is not returnable.
View return policy
Description
KLH Polyclonal specifically detects KLH in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
| KLH | |
| Polyclonal | |
| Biotin | |
| Phosphate Buffered Saline (PBS) containing 0.2% BSA with 0.09% Sodium Azide | |
| Rabbit | |
| Antigen Affinity-purified | |
| Primary | |
| Non-species specific | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1:10000-1:100000, ELISA 1:10000-1:100000 | |
| keyhole limpet hemocyanin (KLH) | |
| KLH was used as the immunogen. | |
| 1 mL | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with Keyhole Limpet Hemocyanin (KLH). | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction