missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody (OTI4F5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70384IR
This item is not returnable.
View return policy
Description
Cadherin-4/R-Cadherin Monoclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cadherin-4/R-Cadherin | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDH4 | |
| Recombinant protein fragment corresponding to amino acids 170-428 of human Cadherin-4/R-Cadherin (NP_001785) produced in E.coli. | |
| 0.1 mL | |
| 1002.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4F5 | |
| Western Blot, Immunohistochemistry | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction