missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody (SR1130), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22545-100ul
This item is not returnable.
View return policy
Description
Cadherin-4/R-Cadherin Monoclonal antibody specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Cadherin-4/R-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1002 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1130 | |
| Western Blot 1:500-1:2000 | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| A synthesized peptide derived from human Cadherin-4/R-Cadherin (Uniprot #: P55283) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction