missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98434CL1
This item is not returnable.
View return policy
Description
Cadherin-8 Polyclonal antibody specifically detects Cadherin-8 in Rat samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| Cadherin-8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| cadherin 8, type 2, cadherin-8, Nbla04261, putative protein product of Nbla04261 | |
| Produced in rabbits immunized with purified, recombinant Rat Cadherin-8 (Accession#: NP_445845.2; Met1-Met621). | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 1006 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction