Learn More
Invitrogen™ CADM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595979
Description
Immunogen sequence: QNLFTKDVTV IEGEVATISC QVNKSDDSVI QLLNPNRQTI YFRDFRPLKD SRFQLLNFSS SELKVSLTNV SISDEGRYFC QLYTDPPQES YTTITVLVPP RNLMIDIQKD TAVEGEEIEV NCTAMASKPA TTIRWFKGNT ELKGKSEVEE WSDMYTVTSQ LMLKVHKEDD GVPVICQVEH PAVTGNLQTQ RYLEVQYKPQ VHIQMTYPLQ GLTREGDALE LTCEAIGKPQ PVMVTWVRVD DEMPQHAVLS GPNLFINNLN KTDNGTYRCE ASNIVGKAHS DYMLYVYDPP TTIPPPTTTT Positive Samples: SW480, Mouse brain, Mouse testis, Mouse liver; Cellular Location: Cell junction, Cell membrane, Single-pass type I membrane protein, synapse.
Mediates homophilic cell-cell adhesion in a Ca (2+)-independent manner. Also mediates heterophilic cell-cell adhesion with CADM3 and PVRL3 in a Ca (2+)-independent manner. Acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. May contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. May act as a synaptic cell adhesion molecule that drives synapse assembly. May be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. May play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.
Specifications
| CADM1 | |
| Polyclonal | |
| Unconjugated | |
| CADM1 | |
| 2900073G06Rik; 3100001I08Rik; AI987920; Bl2; cadm1; Cell adhesion molecule 1; cell adhesion molecule 1 transcript variant 8abc; Igsf4; Igsf4a; Immunoglobulin superfamily member 4; immunoglobulin superfamily member 4A; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4A; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; immunosuperfamily protein Bl2; Necl2; NECL-2; nectin-like 2; nectin-like protein 2; RA175; RA175A; RA175B; RA175C; RA175N; secretory isoform of TSLC1; sgigsf; Spermatogenic immunoglobulin superfamily; ST17; sTSLC-1; Synaptic cell adhesion molecule; SynCAM; SynCam1; truncated CADM1 protein; TSLC1; TSLC-1; TSLC1/Nectin-like 2/IGSF4; tumor suppressor in lung cancer 1; Tumor suppressor in lung cancer 1 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23705, 363058, 54725 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.721 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6AYP5, Q8R5M8, Q9BY67 | |
| CADM1 | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.