Learn More
Invitrogen™ CADM3 Recombinant Rabbit Monoclonal Antibody (3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530127
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CADM3/NECL1/IGSF4B.
Cell adhesion molecule 3 (CADM3), also known as Necl-1, is a gene located on chromosome 1p36.22, encoding a protein that is part of the immunoglobulin superfamily involved in cell-cell adhesion processes. CADM3 is predominantly expressed in the nervous system, particularly in regions such as the brain and spinal cord, where it plays crucial roles in neuronal communication, synapse formation, and maintenance of neural network architecture. The protein functions by mediating homophilic and heterophilic interactions with other cell adhesion molecules, facilitating synapse stabilization and neuronal differentiation. CADM3's activity is essential for proper neural development and function, and dysregulation of its expression has been linked to various neurological disorders. Beyond its role in the nervous system, CADM3 also contributes to tumor suppression in certain cancers, as its expression is often downregulated in tumor tissues. Research into CADM3 continues to explore its potential as a therapeutic target in both neurobiology and oncology.
Specifications
| CADM3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8N126 | |
| Cadm3 | |
| Recombinant Human CADM3 protein (Met1-His330). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 3 | |
| Unconjugated | |
| Cadm3 | |
| BIgR; brain immunoglobulin receptor; CADM3; Cell adhesion molecule 3; dendritic cell nectin-like protein 1 short isoform; IGSF4B; Immunoglobulin superfamily member 4B; immunoglobulin superfamily, member 4B; NECL1; NECL-1; nectin-like 1; nectin-like protein 1; nectin-lke 1; synaptic cell adhesion molecule 3; SYNCAM3; TSLC1-like 1; TSLC1-like protein 1; TSLL1; UNQ225/PRO258 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.