missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CADPS Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77323-0.025mg
This item is not returnable.
View return policy
Description
CADPS Polyclonal specifically detects CADPS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CADPS | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Ca++-dependent secretion activator, CADPS1, Calcium-dependent activator protein for secretion 1, calcium-dependent secretion activator 1, CAPS-1, CAPS1Ca2+-dependent secretion activator, CAPSCa2+-dependent activator protein for secretion, KIAA1121Ca2+-regulated cytoskeletal protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8618 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_899631 | |
| CADPS | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of the human CAPS1. The immunogen is located within the last 50 amino acids of CAPS1. Amino Acid Squence: DSTLNSKTYETIRNRLTVEE | |
| 0.025 mg | |
| Primary | |
| Numerous isoforms of CAPS1 are known to exist; the lower molecular weight bands seen in the immunoblot image are likely to be these isoforms. This CAPS1 antibody is predicted to be specific to CAPS1 and not recognize CAPS2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction