missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAF1 p60 Monoclonal Antibody (SS 53)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA116845
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for CHAF1B (transient overexpression lysate).
Chromatin assembly factor 1 (CAF-1) is a member of the histone chaperone family. It is required for inheritance of epigenetically determined chromosomal states, and in vitro, it promotes the assembly of chromatin during DNA replication and DNA repair. CAF-1 is a complex of three subunits; p150, p60, and p48. This complex is localized to the nucleus of human cells. Both the composition and the biochemical activity of CAF-1 are conserved among human, yeast, and Drosophila.
Specifications
| CAF1 p60 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.1% sodium azide | |
| Q13112 | |
| Chaf1b | |
| Recombinant human CHAF1B. | |
| RUO | |
| 8208 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| SS 53 | |
| Unconjugated | |
| Chaf1b | |
| 2600017H24Rik; C76145; CAF1; CAF-1; CAF1 p60; CAF-1 subunit B; CAF1A; CAF1P60; CAF-I 60 kDa subunit; CAF-I p60; CAF-Ip60; CHAF1B; Chromatin assembly factor 1 subunit B; chromatin assembly factor 1, subunit B; chromatin assembly factor 1, subunit B (p60); chromatin assembly factor I p60 subunit; fe36g06; gef; human chromatin assembly factor-I p60 subunit; LOW QUALITY PROTEIN: chromatin assembly factor 1 subunit B; M-phase phosphoprotein 7; MPHOSPH7; MPP7; p60 CAF1; p60-CAF1; wu:fd07c09; wu:fe36g06; wu:fv38g09; zgc:56096 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction