Learn More
Caffeine-d3 (1-methyl-d3), CDN
Specifications
Specifications
| Chemical Name or Material | Caffeine-d3 |
| CAS | 26351-03-1 |
| Molecular Formula | C8H7D3N4O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i3D3 |
| SMILES | O=C(N(C)C1=C2N(C)C=N1)N(C([2 H])([2 H])[2 H])C2=O |
| IUPAC Name | 3,7-dihydro-3,7-dimethyl-1-(methyl-d3)-1 H-purine-2,6-dione |
| Molecular Weight (g/mol) | 197.2 |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |