missing translation for 'onlineSavingsMsg'
Learn More

Caffeine-d3 (7-methyl-d3), CDN

Product Code. 30316646
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316646 0.1 g 0.1g
30312461 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316646 Supplier CDN Supplier No. CDND59130.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caffeine-d3 (7-methyl-d3)
CAS 26351-04-2
Molecular Formula C8 D3 H7 N4 O2
Quantity 0.1 g
InChI Formula InChI=1 S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3
SMILES [2 H]C([2 H])([2 H])n1cnc2N(C)C(=O)N(C)C(=O)c12
IUPAC Name 1,3-dimethyl-7-(trideuteriomethyl)purine-2,6-dione
Molecular Weight (g/mol) 197.209
Formula Weight 197.0992 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.