Learn More
Caframo™ Pivoting Blade Impeller
GREENER_CHOICE
Product Code. 12937106
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
Product Code. 12937106
Brand: Caframo™ A140
Only null left
Add to basket
Add to basket
Pivoting Blade Impeller with shaft for use with Caframo™ and Fisherbrand™ Overhead Stirrers. Radial flow with moderate shear and moderate vortexing properties.
  • Electro-polished stainless steel 316L
  • Blade dia: 60 mm (2 3/8 in.)
  • Shaft Length: 400 mm
  • Shaft dia: 8 mm (5/16 in)
  • Weight: 126 g (0.3 lb.)
  • Moderate shear with moderate vortexing characteristics
  • Will vortex 5 gallons of water at 800 rpm
  • High speed, Low viscosity mixing, Radial flow pattern
Specifications
Diameter (English) 2.375 in.
Diameter (English) Shaft 0.3125 in.
Diameter (Metric) 60 mm
Diameter (Metric) Shaft 8 mm
For Use With (Equipment) Caframo™ Overhead Stirrer
For Use With (Application) High Speed, Low Viscosity Mixing, Radial Flow Pattern
Length (English) 16 in.
Length (Metric) 400 mm
Type Pivoting Blade Impeller
Weight (Metric) 126 g
Material Electropolished Stainless Steel (316L)
Weight (English) 0.3 lb.