Learn More
Calbindin D-28K Antibody (5A9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50050UV
Product Code. 18685589
  • € 614.00 / 0.10mL
Description
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse
Full-length recombinant human Calbindin D-28K. [UniProt# P05937]