missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 206.00 - € 510.00
Specifications
| Antigen | Calbindin D-28K |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604158
|
Novus Biologicals
NBP2-50028 |
0.1 mL |
€ 510.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669108
|
Novus Biologicals
NBP2-50028SS |
0.025 mL |
€ 206.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calbindin D-28K Polyclonal antibody specifically detects Calbindin D-28K in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry Free-FloatingSpecifications
| Calbindin D-28K | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
| Unconjugated | |
| Chicken | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| Supplied as an aliquot of IgY preparation | |
| CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
| Full-length recombinant human Calbindin D-28K. [UniProt# P05937] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| P05937, P05937 | |
| 793 | |
| IgY | |
| IgY purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title