missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcifediol Monohydrate, Mikromol™
Description
Calcifediol Monohydrate
Specifications
Specifications
| Chemical Name or Material | Calcifediol Monohydrate |
| CAS | 63283-36-3 |
| Analyte or Component Names | Calcifediol Monohydrate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C27 H44 O2 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Calcifediol, Vitamin D3 25-hydroxy monohydrate, 9,10-Secocholesta-5,7,10(19)-triene-3,25-diol, monohydrate, (3beta,5Z,7E)- (9CI) |
| InChI Formula | InChI=1S/C27H44O2.H2O/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29;/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3;1H2/b21-11+,22-12-;/t20-,23+,24-,25+,27-;/m1./s1 |
| SMILES | O.C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C |
| IUPAC Name | (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?