missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcineurin A Recombinant Rabbit Monoclonal Antibody (5I6H8)

Calcineurin A Recombinant Rabbit Monoclonal Antibody (ARC0970)
Brand: Invitrogen™ MA535487
This item is not returnable.
View return policy
Description
Calcineurin A Recombinant Monoclonal Antibody for Western Blot
Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.
Specifications
| Calcineurin A | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P63328, P63329, Q08209 | |
| PPP3CA | |
| A synthetic peptide corresponding to a sequence within amino acids 422-521 of human Calcineurin A (Q08209). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5I6H8 | |
| Unconjugated | |
| PPP3CA | |
| 2900074D19Rik; Ca2+/calmodulin-regulated protein phosphatase; calcineurin; calcineurin A alpha; Calcineurin A1; calcineurin catalytic subunit; calcineurin catalytic subunit delta isoform; Calcineurin subunit A alpha; CalcineurinA; calmodulin-dependent calcineurin A subunit alpha isoform; caln; CALNA; CALNA 1; CALNA1; CAM-PRP catalytic subunit; CCN 1; ccn1; CN; CnA; CNA alpha; CNA1; PP2B alpha 1; PP2BA alpha; PP2CB; ppp2b; ppp3ca; ppp3ca.S; protein phosphatase; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform (calcineurin A alpha); protein phosphatase 3 catalytic subunit alpha; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme S homeolog; protein phosphatase 3, catalytic subunit, gamma isozyme; protein phosphatase-2Ba; serine/threonine protein kinase; serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform; XELAEV_18009418mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19055, 24674, 5530 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction