missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin B Antibody (CALNB/2342), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08464AF350
This item is not returnable.
View return policy
Description
Calcineurin B Monoclonal antibody specifically detects Calcineurin B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Calcineurin B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa) | |
| Recombinant full-length Calcineurin B protein (Uniprot: P63098) | |
| 100 μg | |
| Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway | |
| 5534 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CALNB/2342 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction