missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
€ 398.00 - € 604.00
Specifications
| Antigen | Calcitonin |
|---|---|
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:4000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18419214
|
Novus Biologicals
NB600-1156 |
0.1 mL |
€ 604.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18468543
|
Novus Biologicals
NB600-1156-25ul |
25 μg |
€ 398.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcitonin Polyclonal antibody specifically detects Calcitonin in Human, Mouse, Rat, Gerbil, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Calcitonin | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Sheep | |
| Cancer, Cellular Markers, Neuroscience, Phospho Specific | |
| PBS | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Synthetic peptide corresponding to a portion of rat alpha-calcitonin gene-related peptide (CGRP). | |
| IgG purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Gerbil, Rabbit | |
| P01257 | |
| 796 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title