missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin R Antibody (462802), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4614CL1
This item is not returnable.
View return policy
Description
Calcitonin R Monoclonal antibody specifically detects Calcitonin R in Human samples. It is validated for Flow Cytometry,CyTOF,Gene Knock-Out
Specifications
| Calcitonin R | |
| Monoclonal | |
| CoraFluor 1 | |
| calcitonin receptor, CRT, CTR, CT-R, CTR1 | |
| HEK293 human embryonic kidney cell line transfected with human Calcitonin R, Ala25-Ala474, Accession # NP_001733 | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction | |
| 799 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, CyTOF, Gene Knock-Out | |
| 462802 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction