Learn More
Calcitriol, MedChemExpress
Product Code. 16475487
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Product Code. 16475487
Brand: Medchem Express HY10002
MedChemExpress Calcitriol is the most active metabolite of vitamin D and also a vitamin D receptor (VDR) agonist.
Calcitriol is the most active metabolite of vitamin D and also a vitamin D receptor (VDR) agonist.
Specifications
Chemical Name or Material Calcitriol
CAS 32222-06-3
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 5 mg
Synonym 1,25-Dihydroxyvitamin D3
Solubility Information DMSO : 110 mg/mL (264.02 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (240.02 mM; Need ultrasonic)
SMILES C=C1[C@H](C[C@@H](C/C1=C/C=C2[C@]3([C@@](C)([C@H](CC3)[C@@H](CCCC(C)(O)C)C)CCC/2)[H])O)O
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 96.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only