Learn More
Calcium 2-Oxoglutarate, TRC
Specifications
Specifications
| Chemical Name or Material | Calcium 2-Oxoglutarate |
| CAS | 71686-01-6 |
| Molecular Formula | C5H4O5Ca |
| Quantity | 1 g |
| Synonym | Calcium 2-Oxopentanedioate |
| InChI Formula | InChI=1S/C5H6O5.Ca/c6-3(5(9)10)1-2-4(7)8;/h1-2H2,(H,7,8)(H,9,10);/q;+2/p-2 |
| SMILES | [Ca+2].[O-]C(=O)CCC(=O)C(=O)[O-] |
| IUPAC Name | calcium;2-oxopentanedioate |
| Molecular Weight (g/mol) | 184.16 |
| Formula Weight | 183.9685 |