missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Acetate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Calcium Acetate (AS) |
| CAS | 62-54-4 |
| Molecular Formula | 2 C2 H3 O2 . Ca |
| Quantity | 25 g |
| Synonym | Calcium acetate anhydrous, Acetic acid, calcium salt (2:1), Acetic acid, calcium salt (8CI,9CI), Calcium acetate (6CI), Brown acetate, Calcium diacetate, E 263, Gray acetate, Lime acetate, Lime pyrolignite, PhosLo, Phosex, Royen, Sanopan, Sorbo-Calcion, Teltozan |
| InChI Formula | InChI=1S/2C2H4O2.Ca/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2 |
| SMILES | [Ca+2].CC(=O)[O-].CC(=O)[O-] |
| IUPAC Name | calcium;diacetate |
| Molecular Weight (g/mol) | 158.17 |
| Formula Weight | 157.9892 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction