missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Activated Nucleotidase 1/CANT1 Antibody (OTI7F8) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Calcium Activated Nucleotidase 1/CANT1 Monoclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | Calcium Activated Nucleotidase 1/CANT1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI7F8 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:150 |
| Formulation | Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative |
| Gene Alias | Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1 |
| Gene Symbols | CANT1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?