missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxalate monohydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-3256942
Click to view available options
Quantity:
100 g
500 g
5794-28-5
C2H2CaO5
146.11
MFCD00150039
LQHWSGSWNOHVHO-UHFFFAOYSA-L
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
165350
calcium;oxalate;hydrate
O.[Ca++].[O-]C(=O)C([O-])=O
5794-28-5
C2H2CaO5
146.11
MFCD00150039
LQHWSGSWNOHVHO-UHFFFAOYSA-L
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
165350
calcium;oxalate;hydrate
O.[Ca++].[O-]C(=O)C([O-])=O

Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C
Physical Form Lump/Powder
Quantity 100 g
Assay Percent Range 99%
Solubility Information Slightly soluble in water.
Formula Weight 146.11 (128.10 Anhydrous)
Sensitivity Hygroscopic
Density 2.12
Chemical Name or Material Calcium oxalate, monohydrate
compliance-icons
Product Identifier
  • Calcium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : EDTA, calcium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'