missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium-sensing R/CaSR Antibody (611825) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9608F
This item is not returnable.
View return policy
Description
Calcium-sensing R/CaSR Monoclonal antibody specifically detects Calcium-sensing R/CaSR in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Calcium-sensing R/CaSR | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 611825 | |
| Flow Cytometry, CyTOF-ready | |
| calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441 | |
| Mouse myeloma cell line NS0-derived transfected with recombinant human Calcium-sensing R/CaSR protein, Tyr20-Lys601, Accession # P41180-1 | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction | |
| 846 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction