Learn More
Calcium Standard Solution Concentrate, 100ppm, Reagecon™
BIOPHARMA
Product Code. 11888213 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11888213
Brand: Reagecon 5000802C