Learn More
Invitrogen™ CALCOCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121163
Description
Positive test controls include: HeLa, 293T, MCF7, A-549, Mouse lung, Mouse heart, Mouse thymus. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: LVQLSESKRE LTELRSALRV LQKEKEQLQE EKQELLEYMR KLEARLEKVA DEKWNEDATT EDEEAAVGLS CPAALTDSED ESPEDMRLPP YGLCERGDPG SSPAGPREAS PLVVISQPAP ISPHLSGPAE DSSSDSEAED EKSVLMAAVQ SGGEEANLLL PELGSAFYDM ASGFTVGTLS ETSTGGPATP TWKECPICKE RFPAESDKDA LEDHMDGHFF FSTQDPFTFE.
CALCOCO1 (calcium-binding and coiled-coil domain-containing protein 1), also known as cocoa, calphoglin, sarcoma antigen NY-SAR-3 or coiled-coil coactivator protein, is a 691 amino acid protein that shuttles between the cytoplasm and nucleus and functions as coactivator for aryl hydrocarbon and nuclear receptors. A member of the CALCOCO family, CALCOCO1 is forms a calphoglin complex with PPA1 and PGM 1 and contains multiple functional domains through which it acts as a component of both the androgen signaling pathway and the Wnt/beta-catenin signaling pathway.
Specifications
| CALCOCO1 | |
| Polyclonal | |
| Unconjugated | |
| Calcoco1 | |
| 1810009B06Rik; calcium binding and coiled coil domain 1; calcium binding and coiled-coil domain 1; calcium-binding and coiled-coil domain-containing protein 1; Calcoco1; Calphoglin; Cocoa; Coiled-coil coactivator protein; coiled-coil leucine zipper coactivator 1; coiled-coil transcriptional coactivator; Gcap11; granule cell antiserum positive 11; GRIP1-interacting protein; inorganic pyrophosphatase activator; KIAA1536; MGC128949 protein; mKIAA1536; PP13275; Sarcoma antigen NY-SAR-3; UNQ2436/PRO4996 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57658, 67488 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CGU1, Q9P1Z2 | |
| Calcoco1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 462-691 of human CALCOCO1 (NP_065949.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.