missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALCOCO2 Monoclonal Antibody (OTI7A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502106
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.Specifications
| CALCOCO2 | |
| Monoclonal | |
| 0.92 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13137 | |
| CALCOCO2 | |
| Full length human recombit protein of human CALCOCO2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI7A7 | |
| Unconjugated | |
| CALCOCO2 | |
| NDP52 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10241 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction