Learn More
CALCOCO2 Monoclonal Antibody (OTI7A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502106S
Encompass_Preferred
 View more versions of this product
Product Code. 17596001
  • € 242.00 / 30µL
Description
CALCOCO2 Monoclonal antibody specifically detects CALCOCO2 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
CALCOCO2
Monoclonal
0.92 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UJM8
Mouse
Affinity chromatography
RUO
311446, 485774, 54363
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Flow Cytometry, Immunocytochemistry, Western Blot
OTI7A7
Unconjugated
HAO1
2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974
Full length human recombinant protein of human HAO1 produced in HEK293T cell.
30 μL
Primary
Canine, Human, Rat