Learn More
Invitrogen™ CALCOCO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530367
Description
Recommended positive controls: Raji, HepG2, HepG2 whole cell extract (untreated), HepG2 whole cell extract (3 μM Thapsigargin treatment for 12 hr), HepG2 whole cell extract (3 μM Thapsigargin treatment for 24 hr), Huh-7 (untreated), Huh-7 (3 μM Thapsigargin treatment for 12 hr), Huh-7 (3 μM Thapsigargin treatment for 24 hr). Predicted reactivity: Human (99%), Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
Specifications
| CALCOCO2 | |
| Polyclonal | |
| Unconjugated | |
| CALCOCO2 | |
| 2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10241, 76815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| A2A6M5, Q13137 | |
| CALCOCO2 | |
| Recombinant protein encompassing a sequence within the center region of human NDP52. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.