missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CALCOCO2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16383865
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16383865 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16383865 Supplier Invitrogen™ Supplier No. PA587007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALCOCO2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CALCOCO2
Gene Accession No. A2A6M5, Q13137
Gene Alias 2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974
Gene Symbols CALCOCO2
Host Species Rabbit
Immunogen Recombinant fusion protein of human NDP52.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10241, 303479, 76815
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.