missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALCRL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111234
This item is not returnable.
View return policy
Description
Immunogen sequence: RNWNQYKIQF GNSFSNSEAL RSASYTVSTI SDGPGYSHDC PSEHLNGKSI HDIENVL Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.
CRLR is a receptor for calcitonin gene-related peptide type 1. The activity of CRLR is mediated by G proteins which activate adenylyl cyclase. CRLR expression has been reported in brain, lung, blood vessel, liver, and intestinal tract. ESTs have been isolated from B-cell/lung/testis, bome marrow, embryo, lung and synovium libraries.
Specifications
| CALCRL | |
| Polyclonal | |
| Unconjugated | |
| CALCRL | |
| AV071593; Calcitonin gene-related peptide type 1 receptor; calcitonin receptor like receptor; calcitonin receptor-like; calcitonin receptor-like receptor; CALCRL; CGRP receptor; CGRP type 1 receptor; Cgrp1 receptor; CGRPR; CLR; CRLR; RATCRLR; RNCLR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10203 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16602 | |
| CALCRL | |
| Recombinant Protein corresonding to Human CALCRL. Recombinant protein control fragment (Product #RP-106121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction