missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcyclin Binding Protein Mouse Anti-Human, Clone: 2E3, Abnova&Trade;
Mouse Monoclonal Antibody
Brand: Abnova H00027101-M01.100ug
This item is not returnable.
View return policy
Description
calcyclin binding protein Monoclonal antibody specifically detects calcyclin binding protein in Human samples. It is validated for ELISA, Western Blot, Western Blot, In situ PLASpecifications
| calcyclin binding protein | |
| Monoclonal | |
| Unconjugated | |
| BC005975 | |
| Mouse | |
| 100 μg | |
| MASEELQKDLEEVKVLLEKATRKRVRDALTAEKSKIETEIKNKMQQKSQKKAELLDNEKPAAVVAPITTGYTVKISNYGWDQSDKFVKIYITLTGVHQVPTENVQVHFTERSFDLLVKNLNGKSYSMIVNNLLKPISVEGSSKKVKTDTVLILCRKKVENTRWDYLTQVEKECKEKEKPSYDTETDPSEGLMNVLKKIYEDGDDDMKRTINKAWVESREKQAKGDTEF | |
| 27101 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot, In situ PLA | |
| 2E3 | |
| In 1x PBS, pH 7.4 | |
| GIG5/MGC87971/PNAS-107/RP1-102G20.6/S100A6BP/SIP | |
| CACYBP (AAH05975, 1 a.a. ∽ 228 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction