missing translation for 'onlineSavingsMsg'
Learn More

Caldesmon/CALD1 Antibody (CALD1/820 + h-CALD), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-47819V

Product Code. 18109648

  • € 611.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Caldesmon/CALD1
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CALD1
Recombinant full-length human Caldesmon/CALD1 protein (Caldesmon/CALD1/820); Crude human uterus extract (h-CALD) (Uniprot: Q05682)
0.1 mL
Primary
Recognizes a protein of 150kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120'150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70'80kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This monoclonal antibody recognizes only the 150kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.
Store at 4C in the dark.
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
CALD1/820 + h-CALD
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Mouse
Protein A or G purified
Cytoskeleton Markers, Phospho Specific
800
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.